Transgenic Dough Diet™ for Rodents, Sterile, Bio-Serv

Supplier: Bio-Serv
S3472
89067-442EA 54.84 USD
89067-442
Transgenic Dough Diet™ for Rodents, Sterile, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete, soft diet with higher protein and fat levels to comply with NRC and Transgenic-users' recommendations

  • Designed for rodents with chewing, dental or mobility impairments
  • Cookie dough consistency replaces labor intensive, nutrient-diluted mush diets
  • Ideal for recovering rodents and weanlings or any time a soft diet is needed
  • Rich bacon flavor stimulates appetite in dogs and rodents
  • Place in feeder cup or on cage floor
  • Sterilized by gamma irradiation
  • Nutritionally assayed
  • Shelf Life: 6 months refrigerated, 12 months frozen

Ingredients: Sucrose, Whole Milk Powder, Casein, Soybean Oil, Cellulose, Corn Syrup, Vitamin Mix, Mineral Mix, Glycerin, Bacon Flavor, DL-Methionine, Choline Bitartrate, Phenylalanine, tBHQ.

Caloric value: 3.83 kcal/g
Nutritional ash content: 3.3%
Nutritional carbohydrate content: 46.5%
Nutritional fat content: 12.4%
Nutritional fibre content: 4.8%
Nutritional moisture content: <12%
Nutritional protein content: 21.2%
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR