Timothy Hay Cubes, Certified, Bio-Serv

Supplier: Bio-Serv
F1011
89067-422EA 108.97 USD
89067-422
Timothy Hay Cubes, Certified, Bio-Serv
Animal Supplies Animal Feed
Contains 100% sun-cured timothy hay, steam pressed

  • No binders added
  • Excellent source of fiber
  • Nutritionally assayed.
  • Shelf Life: 24 months, cool dry storage.

Caloric value: 1.91 kcal/g
Nutritional ash content: 8.5%
Nutritional carbohydrate content: 31.5%
Nutritional fat content: 1.0%
Nutritional fibre content: 35.0%
Nutritional moisture content: <10%
Nutritional protein content: 14.0%

Certifications: Certified (Contaminant Screened).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR