Rabbit Stix™, Certified, Bio-Serv

Supplier: Bio-Serv
F2831
89067-420EA 145.65 USD
89067-420
Rabbit Stix™, Certified, Bio-Serv
Animal Supplies Animal Feed
High-fiber, pineapple-flavored maintenance diet, supplement or enrichment treat.

  • Contains papain, commonly used to prevent hairballs, and is an excellent source of Vitamin C
  • Use as an appetite stimulant for new arrivals, post-op or for debilitated conditions
  • Also recommended for Guinea Pigs
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Alfalfa Meal, Ground Corn, Wheat Middlings, Dehulled Soybean Meal, Ground Oats, Ground Soybean Hulls, Cane Molasses, Soy Protein Isolate, Soybean Oil, Mineral Mix, Vitamin Mix, Choline Chloride, Lubricant, Papain, Ascorbic Acid, Pineapple Flavor, Niacin.

Caloric value: 2.73 kcal/g
Nutritional ash content: 7.6%
Nutritional carbohydrate content: 42.0%
Nutritional fat content: 3.7%
Nutritional fibre content: 22.0%
Nutritional moisture content: <10%
Nutritional protein content: 18.0%

Certifications: Certified (Contaminant Screened).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR