Softies™ Primate Diet, Certified, Bio-Serv

Supplier: Bio-Serv
F3478
89067-392EA 269.23 USD
89067-392
Softies™ Primate Diet, Certified, Bio-Serv
Animal Supplies Animal Feed
Soft, pelleted, nutritionally complete diet is ideal for chewing difficulties, post-op care or finicky eaters

  • Excellent transition diet for weanlings
  • Nutritionally assayed
  • Can be used with other species
  • Shelf Life: 6 months refrigerated

Ingredients: Sucrose, Casein, Whole Milk Powder, Soy Flour, Corn Syrup Solids, Cellulose, Corn Oil, Molasses, Mineral Mix, Wheat Bran, Magnesium Stearate, Acacia Gum, Glycerin, Vitamin Mix, Natural and Artificial Flavors, Choline Chloride, DL-Methionine, Ascorbic Acid, Vitamin E Acetate, BHA, Food Dye.

Caloric value: 3.63 kcal/g
Nutritional ash content: 5.3%
Nutritional carbohydrate content: 44.6%
Nutritional fat content: 10.4%
Nutritional fibre content: 8.5%
Nutritional moisture content: <10%
Nutritional protein content: 22.9%
Vitamin C: 2.7 mg/g
Vitamin D3: 3.6 IU/g
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR