Primilac™ Liquid Infant Primate Diet, Bio-Sev

Supplier: Bio-Serv
F0554SP
89067-386EA 91.39 USD
89067-386
Primilac™ Liquid Infant Primate Diet, Bio-Sev
Animal Supplies Animal Feed
A milk substitute for infant non-human primates containing selenium, chromium and fluoride, which are not added to human formulas.

  • Contains appropriate amounts of protein, fat and carbohydrates for most primates
  • Contains a higher level of Vitamin C per serving compared to human formulas
  • Easy to prepare; no blender needed. Add water, shake and pour. Use with Bio-Serv's 160 oz. Shake and Pour Bottle or your own mixing vessel.
  • Nutritionally assayed

Shelf Life: 6 months refrigerated. If prepared, refrigerate unused portion up to 3 days. Prepared diet should not be kept more than 48 hours at room temperature.

Ingredients: Lactose, Casein, Maltodextrin, Coconut Oil, Corn Oil, Chicken Fat, Mineral Mix, Cellulose, Lecithin, Suspending Aids, Vitamin Mix, Choline Dihydrogen Citrate, Banana Flavor, L-Cystine, BHT.

Nutritional carbohydrate content: 54.5 kcals/6 oz.
Nutritional fat content: 67.8 kcals/6 oz.
Nutritional protein content: 22.5 kcals/6 oz.
Total caloric profile: 144.7 kcals/6 oz.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR