Fruit and Nut Foraging Mix, Certified, Bio-Serv

Supplier: Bio-Serv
F3084-5
89067-313EA 118.74 USD
89067-313
Fruit and Nut Foraging Mix, Certified, Bio-Serv
Animal Supplies Animal Feed
Delicious, natural foraging mixture.

  • Use with Tuff Turf Foraging Board or scatter into bedding
  • Contains no shells
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Peanuts, Sunflower Seeds, Banana Chips and Raisins.

Caloric value: 5.26 kcal/g
Nutritional ash content: 2.6%
Nutritional carbohydrate content: 32.4%
Nutritional fat content: 36.0%
Nutritional fibre content: 3.0%
Nutritional moisture content:<10%
Nutritional protein content: 18.0%

Certifications: Certified (Contaminant Screened).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR