Beef-Flavored Treats, Certified, Bio-Serv

Supplier: Bio-Serv
F3736 F05357
89067-250EA 222.69 USD
89067-250 89067-252
Beef-Flavored Treats, Certified, Bio-Serv
Animal Supplies Animal Feed
Nutritionally fortified, roast beef-flavored treats.

  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.

Caloric value: 3.39 kcal/g for Beefy Bites™, 118.30 kcal/Block for Beefy Blocks™
Nutritional ash content: 7.6% for Beefy Bites™, 7.9% for Beefy Blocks™
Nutritional carbohydrate content: 54.8% for Beefy Bites™, 54.0% for Beefy Blocks™
Nutritional fat content: 3.8%
Nutritional fibre content: 3.8% for Beefy Bites™, 3.9% for Beefy Blocks™
Nutritional moisture content: <10%
Nutritional protein content: 21.5% for Beefy Bites™, 21.9% for Beefy Blocks™

Certifications: Certified (Contaminant Screened).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR