Pliers, Round Nose, Excelta
Supplier: Excelta
2843 2843L 743 2643 2843-CR 2843SL
63041-718EA 39.81 USD
63041-718 63041-800 63041-802 89067-242 76242-520 76246-966 76471-598 76471-600
Pliers, Round Nose, Excelta
Pliers
Round nose pliers are great for creating loops in wire with consistant radius.
  • Round nose for forming wires
  • Ergonomic handle, dual leaf springs
  • Stainless steel construction-corrosion resistance
  • Different sizes depending on application
Round nose pliers are made of AISI 440 stainless steel (with the exception of the 743) and include two leaf springs for more ergonomic control. They are offered in a small or medium, depending on the application. They are most commonly used to form wires.