SHOWA NSK24 Nitrile Supported, Chemical Resistant, 14 Glove, Showa
Supplier: Showa
NSK24-10 NSK24-08
80090-034CS 1257.67 USD
80090-034 89065-416
SHOWA NSK24 Nitrile Supported, Chemical Resistant, 14 Glove, Showa
Gloves
Double dipped Nitrile coating and rough finish provide excellent grip, and protection from chemicals.
  • Rough grip
  • 14" length
  • Highly flexible coating
These gloves are used to handle a wide array of solvents, oils, and acids. Double dipped coating ensures added protection when working with hazardous chemicals.