IVT1000CR Cleanroom Vacuum with Safe-Pak and Accessory Kit, Nilfisk
Supplier: Nilfisk
01798180
89065-246EA 5212.69 USD
89065-246
IVT1000CR Cleanroom Vacuum with Safe-Pak and Accessory Kit, Nilfisk
Vacuum Cleaners
The Nilfisk IVT1000CR is a cleanroom compliant vacuum, approved for cleanrooms up to ISO4 (Class 10).
  • Compact design
  • ULPA exhaust filter standard
  • Trolley is autoclaveable for sterilization
  • Locking clamps ensure airtight sealing
  • Modular construction makes it easy to clean and service
Ordering information: Accessories included are plastic hose, plastic cuff, curved stainless steel wand, straight stainless steel wand, hose twist plug, floor nozzle, dust brush, and rubber crevice nozzle. Available with the Nilfisk disposable HEPA-filtered SAFE-PAK container for safe collection, containment and disposal of potent compounds. Featuring multistage filtration, AISI316 stainless steel construction and a quiet 61 dB(A) noise level.
Packaging: Accessories could add additional weight and boxes to the order.