Adapter, 75 Degree Angle, Ace Glass Incorporated
Supplier: Ace Glass
5090-10 5090-15 5090-35 9077-02 9077-06 9077-16
89051-310EA 121.2 USD
89051-310 89051-312 89051-314 89064-838 89064-840 89064-842
Adapter, 75 Degree Angle, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features standard taper or spherical joints at the bottom and side with a thermometer joint at the top.
  • 75° angle sidearm
  • Bottom and side [ST] or spherical joints
  • Top [ST] outer thermometer joint
  • Fabricated from borosilicate glass
Adapter comes with standard taper or spherical joints at the bottom and side. At the top is a standard taper 10/30 outer thermometer joint.