Mini #7 Ace-Thred with Hose Connection Adapter, Ace Glass Incorporated
Supplier: Ace Glass
5261-20 5261-16 5261-06 5261-08 5261-12
89051-406EA 85.15 USD
89051-406 89051-498 89064-824 89064-826 89064-828
Mini #7 Ace-Thred with Hose Connection Adapter, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This "Mini" adapter has a standard taper inner joint at the bottom, #7 Ace-Thred at the top and a serrated hose connection.
  • Ace-Thred at top
  • Bottom [ST] inner joint
  • Serrated hose connection
  • Fabricated from borosilicate glass
Adapter will accommodate thermometers, and bleed tubes up to 7mm in diameter. Suitable for most vacuum work.
Ordering information: Supplied complete with 89051-122 nylon bushing and 89060-958 FETFE o-ring.