Beaded Pipe to Conical Pipe Coupling Assembly, Ace Glass Incorporated
Supplier: Ace Glass
8866-03 8866-05 8866-07 8866-09
89064-696EA 787.07 USD
89064-696 89064-698 89064-700 89064-702
Beaded Pipe to Conical Pipe Coupling Assembly, Ace Glass Incorporated
Clamps General Purpose Clamps
To join beaded end pipe to conical end pipe
Ordering information: Kit includes two flanges, inserts, bolts and TFE sheath gasket.