Glass Check Valve with Ground Seat, Ace Glass Incorporated
Supplier: Ace Glass
8774-09
89064-042EA 134.29 USD
89064-042
Glass Check Valve with Ground Seat, Ace Glass Incorporated
Valves Vacuum Valves
All-glass check valve with ground seat and precision ground solid ball
Suitable with mercury in the vertical position, more sensitive to other fluids in the horizontal position. Connections are 8 mm O.D.