Vacuum Trap Assembly, Ace-Thred Top and Inlet/Outlet, Easy-Action Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
8748-47
89063-890EA 2120.76 USD
89063-890
Vacuum Trap Assembly, Ace-Thred Top and Inlet/Outlet, Easy-Action Stopcock, Ace Glass Incorporated
Vacuum Traps
This twin chambered dry ice vacuum trap is designed to be used with rotary evaporators or any other large solvent evaporation applications
The inlet/outlet ports have been modified with #15 Ace-Threds for use with PTFE "Ace-Safe" tubing connectors to make an easy, leak-tight, safe connect/disconnect to tubing. These ports are located on the sides of chambers; one is for connection to the evaporator, the other to the vacuum source. Both are for use with 1/2-inch I.D. tubing. The stopcock at bottom is 0-5mm Easy-Action threaded style with double PTFE ring seals on plug, for draining both chambers simultaneously. Normally, the condensate will collect in the chamber connected nearest to the evaporator until it is filled to crossover between chambers. Then the second chamber will fill. The capacity of the vacuum traps is 450mL each. The capacity of the chambers with the traps inserted is approximately 600mL each. Overall height of the unit is approximately 20 inches.
The unique, squatty design is 18 inches high, and the use of twin chambers offers increased condensing capacity that would be attained only with a much taller, single chambered condenser. The vacuum traps are secured into the main body chambers via #50 Ace-Threds using bushings and o-rings. This type of connection allows for easy removal for cleaning. Fabricated from borosilicate glass.