Temperature Measurement Apparatus for Hydrogenation/Gas Apparatus, Ace Glass Incorporated

Supplier: Ace Glass
8648-57
89063-384EA 1445.6 USD
89063-384
Temperature Measurement Apparatus for Hydrogenation/Gas Apparatus, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This borosilicate glass vessel, offset adapter, coupling and modified top bottle clamp are used with shaker type Parr Hydrogenation/Gas Apparatus I (89058-912) to allow temperature measurement while connected to hydrogen source

For use with three larger vessels only, with #25 Ace-Thred neck.

Ordering information: This complete apparatus comes complete with three borosilicate glass reaction vessels (89063-232, 89063-238, 89063-240), offset adapter (89051-324) with two #7 and one #15 Ace-Thred connections, #7 Ace-Thred nylon bushing with o-ring (89051-128), PTFE coupling (89052-672) #15More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR