Pressure Tubes with Ace-Thred and Back Seal Plugs, Ace Glass
Supplier: Ace Glass
8648-03 8648-06 8648-08 8648-103 8648-106 8648-110 8648-114 8648-116 8648-118 8648-122 8648-125 8648-128 8648-137 8648-246 8648-248 8648-250 8648-252 8648-254 8648-309 8648-311 8648-313 8648-89 8648-97 8648-98 8648-130
89063-160EA 140.11 USD
89063-160 89063-164 89063-168 89063-176 89063-182 89063-190 89063-196 89063-200 89063-204 89063-212 89063-218 89063-224 89063-236 89063-308 89063-312 89063-318 89063-322 89063-326 89063-338 89063-342 89063-346 89063-424 89063-428 89063-430 76318-588
Pressure Tubes with Ace-Thred and Back Seal Plugs, Ace Glass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Heavy-wall, borosilicate glass tubes are designed for polymerization and analysis of halogens by thermal decomposition in the presence of lime
Hydrogenation bottles have a flat bottom and a lower recommended operating pressure of 60psi. Bottle pressure ratings are determined at ambient temperature.
Tubes feature Ace-Thred closures which allow for quick and simple plug sealing and removal. Supplied with back seal plug and O-ring, assuring a tight and secure seal without the need for any extra equipment. Closures are reusable. Tubes are available in multiple body diameters and lengths, with #15, #25, or #36 Ace-Thred. Tube length is measured below the thread. Tube pressure ratings are determined at 120°C (248°F). Each tube is pressure tested at 1.5 times the rating.
Ordering information: Each tube is supplied with a PTFE back seal plug and FETFE O-ring. Replacement O-rings and O-rings of different composition are available. For monitoring purposes, tube can be assembled with a stainless steel pressure gauge (0–160psi), using an optional adapter. In addition, stainless steel tube extensions are available to allow use of tube in a furnace. Tubes can be fabricated in other lengths and diameters. For pressure work, a front seal plug is recommended; see 89063-172 series for more information. For replacement tubes and bottles, see 89063-208 series.