Round-Head Glass Stoppers, Ace Glass Incorporated
Supplier: Ace Glass
8250-02 8250-04 8250-06 8250-08 8250-10 8250-12 8250-14 8250-16 8250-20 8250-24 8251-02 8251-06 8251-08 8251-12 8251-16 8255-04 8255-06 8255-08 8255-12 8255-14 8255-16 8255-18 8255-20 8260-04 8260-06 8260-08 8260-10 8260-12 8260-14 8260-16 8260-18 8250-28 8255-10 8255-114 8255-124 8255-145
89062-814EA 31.95 USD
89062-814 89062-816 89062-818 89062-820 89062-822 89062-824 89062-826 89062-828 89062-830 89062-832 89062-834 89062-836 89062-838 89062-840 89062-842 89062-844 89062-846 89062-848 89062-852 89062-854 89062-856 89062-858 89062-860 89062-862 89062-864 89062-866 89062-868 89062-870 89062-872 89062-874 89062-876 89084-702 10099-498 10099-500 10099-502 10099-504
Round-Head Glass Stoppers, Ace Glass Incorporated
Stoppers Glass Stoppers
Round-head style glass stoppers are available in a variety of lengths and sizes
Stoppers are available in hollow or solid types. All stoppers are manufactured from borosilicate glass.