Stopcock Plugs, Metering Valve, 1:5 PTFE, Ace Glass Incorporated
Supplier: Ace Glass
8232-14 8232-16
89062-806EA 128.47 USD
89062-806 89062-808
Stopcock Plugs, Metering Valve, 1:5 PTFE, Ace Glass Incorporated
Stopcocks
Stopcock plug 89062-806 is designed for use with Retainer O-Ring size –011
Stopcock plug 89062-808 is designed for use with Retainer O-Ring size –111.