Stopcock Plugs, Straight Bore, 1:5 PTFE, Ace Glass Incorporated
Supplier: Ace Glass
8224-04 8224-08 8224-12 8224-16 8224-18
89062-770EA 35.06 USD
89062-770 89062-772 89062-774 89062-776 89062-778
Stopcock Plugs, Straight Bore, 1:5 PTFE, Ace Glass Incorporated
Stopcocks
Stopcock plug 89062-770 is designed for use with Retainer O-Ring size –011
Stopcock plugs 89062-772, -774, and -776 are designed for use with Retainer O-Ring size –111. Stopcock plug 89062-778 is designed for use with Retainer O-Ring size –208.