Newman Type Stopcock, Ace Glass
Supplier: Ace Glass
8152-04 8152-06
89062-374EA 217.82 USD
89062-374 89062-376
Newman Type Stopcock, Ace Glass
Stopcocks
Used with distilling head to enable operator to determine reflux ratios and take-offs.