Hi-Lube Heavy-Duty Liquid Stirrer Lubricant, Ace Glass Incorporated
Supplier: Ace Glass
8119-07
89062-230EA 62.62 USD
89062-230
Hi-Lube Heavy-Duty Liquid Stirrer Lubricant, Ace Glass Incorporated
Lubricants
Chlorofluorocarbon grease is designed for use at speeds up to 6000rpm with Ace Glass standard glass assemblies.
This lubricant has a high chemical inertness, and is unaffected by strong acids and alkalis. Soluble in most organic solvents. It is sitable for use with oxidizing gases. It also has high heat resistance, and is thermally stable up to 260°C (500°F). Non-flammable; does not carbonize on decomposition. Available in convenient one ounce (30mL) size.
Ordering information: For long stirring periods (over one hour), the use of a water-cooled type bearing is recommended. For information on other uses, please contact your Avantor representative.