Adapter, Lubricant Trap, High-Vacuum, 10mm, Ace Glass Incorporated
Supplier: Ace Glass
8113-10 8113-20 8113-25
89062-214EA 161.44 USD
89062-214 89062-216 89062-218
Adapter, Lubricant Trap, High-Vacuum, 10mm, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This trap adapter has an inner tube through which the stirring shaft passes, and it, plus the included O-Ring, prevents the lubricant (or foreign particles) from contaminating the flask contents.
  • PTFE O-Ring supplied with each unit
  • Manufactured from borosilicate glass
This lubricant trap adapter is primarily designed for use with bearings like 89061-802.
Ordering information: Replacement O-Rings are available only via special order. Contact your Avantor representative for details.