Stirrer Blades, Banana Type, PTFE, 10mm, Ace Glass Incorporated
Supplier: Ace Glass
8087-05 8087-07 8087-09 8087-11 8087-13 8087-15 8087-19
89062-104EA 58.9 USD
89062-104 89062-106 89062-108 89062-110 89062-112 89062-114 89062-116
Stirrer Blades, Banana Type, PTFE, 10mm, Ace Glass Incorporated
Stirrers Stirring Blades
These PTFE, banana shaped stirrer blades are designed for use with 10mm O.D
button-type stir shafts. The 3mm thick blades have a number of perforations, and are designed to closely fit various sizes of round bottom flasks.