High Vacuum Bearing, Trubore®, Ace Glass Incorporated
Supplier: Ace Glass
8055-10
89061-814EA 312.27 USD
89061-814
High Vacuum Bearing, Trubore®, Ace Glass Incorporated
Stirrers Stirring Tools
This bearing is the standard liquid seal type except that the use of Trubore® tubing enables this unit to perform very satisfactorily under high vacuum conditions
I.D. is 10 mm. For use with 10 mm shafts.