Trubore® Bearing, Straight, Ace Glass Incorporated
Supplier: Ace Glass
8036-10
89061-690EA 88.46 USD
89061-690
Trubore® Bearing, Straight, Ace Glass Incorporated
Stirrers Stirring Tools
Interchangeable bearing is designed to be used with precision ground stirring shafts.
  • Recommended top stirring speed, when used with appropriate stirring lubrication is 1500 rpm
  • Manufactured from borosilicate glass