Vacuum Sublimation Apparatus, Large Scale, Ace Glass Incorporated
Supplier: Ace Glass
8027-02 8027-05 8027-07 8027-09 8027-14 8027-25
89061-654EA 864.41 USD
89061-654 89061-656 89061-658 89061-660 89061-662 89061-664
Vacuum Sublimation Apparatus, Large Scale, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This large-scale 2L sublimation apparatus is constructed of heavy wall glass.
  • Unique design offers both a sublimation apparatus as well as a two-liter reaction kettle
  • Unit will sublime approximately 100 gms of solid sample
  • Constructed of heavy-wall glass
Unit will sublime approximately 100 gms of solid sample. Unique design offers both a sublimation apparatus as well as a two-liter reaction kettle. Cold finger tube has a standard taper 34/35 joint which positions in the center neck and is held by means of a plastic bushing to a 300 mL condensate flask. The wide flange opening of the kettle allows for easy removal and access to the condenser flask. Side joints are standard taper 24/40. Kettle flange has an o-ring grove for a CAPFE O-Ring, (supplied) for easy grease free seal. Head and kettle are joined with a quick release clamp.
Ordering information: Complete system has all the components listed below. 89061-662 stopcock adapter uses size "C" (⅜") hose connections. 89061-658 condenser tube uses size A (5/16") hose connections.