Clamps, Spherical Joint, Union Type, Ace Glass Incorporated
Supplier: Ace Glass
7666-05 7666-10 7666-15 7666-20 7666-30
89060-228EA 101.79 USD
89060-228 89060-230 89060-232 89060-234 89060-236
Clamps, Spherical Joint, Union Type, Ace Glass Incorporated
Clamps Standard Taper and Spherical Joint Clamps
These spherical joint, union-type clamps are fabricated from aluminum
This type of clamp affords a positive, uniform clamping pressure. The clamp threads together and covers the entire joint.