Joints with O-Ring Seal, Ace Glass Incorporated
Supplier: Ace Glass
7646-02 7646-04 7646-06 7646-08 7646-10 7646-12 7646-14 7646-16 7646-18
89059-970EA 21.38 USD
89059-970 89059-972 89059-974 89059-976 89059-978 89059-980 89059-982 89059-984 89059-986
Joints with O-Ring Seal, Ace Glass Incorporated
These joints are tooled with a groove to accommodate an O-Ring and provide a greaseless, vacuum-tight connection.
  • O-ring included
  • Vacuum-tight connection
  • Manufactured from borosilicate glass
Adapter joints are held together with pinch-type clamps (see table below for details).
O-ring is made of FETFE. FETFE, an Ace Glass exclusive compound, is a fluoroelastomer with TFE additives. It offers good compression set along with excellent temperature (to 230°C) and chemical compatibility. USP Class VI material.
Ordering information: Supplied complete with FETFE O-Ring.