Inner Joints, Extended Drip Tip, Ace Glass Incorporated
Supplier: Ace Glass
7640-02 7640-06 7640-08 7640-10 7640-12 7640-14
89059-844EA 37.59 USD
89059-844 89059-846 89059-848 89059-850 89059-852 89059-854
Inner Joints, Extended Drip Tip, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This is a standard taper joint with lower reduced tube.
  • [ST] joint
  • Lower reduced tube
The lower tube is reduced in diameter to size listed. The length of the extended tube is 101 mm.