Inner Joints, with Reduced Extending Tube, Ace Glass Incorporated
Supplier: Ace Glass
7635-05 7635-10
89059-838EA 45.57 USD
89059-838 89059-840
Inner Joints, with Reduced Extending Tube, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This is a standard taper inner joint with extending tube reduced in diameter.
  • [ST] inner joint
  • Reduced extending tube
Adapters feature inner joints with reduced ends.