Outer Joints, Microscale, Threaded, Ace Glass Incorporated
Supplier: Ace Glass
7609-05 7609-07 7609-08 7609-09 7609-15 7609-17
89059-670EA 30.07 USD
89059-670 89059-672 89059-674 89059-676 89059-678 89059-680
Outer Joints, Microscale, Threaded, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
These outer ground standard taper joints with external thread are used for making an “O-Ring-CAP SEAL” connection with mating inner joint.
  • Forms an O-ring–cap seal connection
  • Cap and O-ring not included
Used in fabricating ACE Microscale glassware. Joints not supplied with cap or O-Ring.