Pressure Filtering Tube, Ace Glass Incorporated
Supplier: Ace Glass
7208-08 7208-12 7208-02
89058-448EA 143.36 USD
89058-448 89058-450 89058-476
Pressure Filtering Tube, Ace Glass Incorporated
Funnels Filtration Funnels Filter Funnels
Tube is designed for use in filtering glucose, agar, and serums
Equipped with an integral glass frit, which is available in two different porosities. The top accepts a #3 or #4 rubber stopper. Fabricated from borosilicate glass.