Erlenmeyer Flask, [ST Joint], Ace Glass Incorporated
Supplier: Ace Glass
6965-26 6965-20 6965-22 6971-24 6965-25 6965-27 6965-28 6965-38 6965-40 6965-41 6971-42 6965-61 6965-62 6965-63 6965-64 6971-26 6965-24 6971-27 6965-42 6971-44 9471-02 9471-04 9471-10 9471-06 9471-12 9471-08
89057-298EA 65.65 USD
89057-298 89057-302 89057-304 89057-306 89057-308 89057-312 89057-314 89057-316 89057-318 89057-320 89057-322 89057-324 89057-326 89057-328 89057-330 89057-336 89057-344 89057-348 89057-350 89057-352 89074-646 89074-648 89074-770 89074-836 89074-862 89074-916
Erlenmeyer Flask, [ST Joint], Ace Glass Incorporated
Flasks Erlenmeyer Flasks
Erlenmeyer flask with a reinforced (on full sized joints only) standard taper outer joint.
  • Fabricated from borosilicate glass
  • Some sizes available with heavy walls
Heavy wall flasks are approximately 30% heavier than standard wall flasks.