Adapter, Freeze Drying, Wide Neck, Ace Glass
Supplier: Ace Glass
6696-50
89055-792EA 189.62 USD
89055-792
Adapter, Freeze Drying, Wide Neck, Ace Glass
Adapters Adapters, Joints with Tube Connectors
This wide-neck connecting adapter is used to connect freeze drying vessels with 48 mm O.D.
necks to #25 Ace-Threds on 89055-766 "umbrella" manifold. The O.D. of the straight side is 25 mm. Fits a 89059-230 nylon #50 Ace-Thred bushing. Fabricated from borosilicate glass.