Distillation Receiver, Jacketed, Graduated, Ace Glass
Supplier: Ace Glass
6642-08
89055-740EA 2399.31 USD
89055-740
Distillation Receiver, Jacketed, Graduated, Ace Glass
Distillation Components Distillation Receivers
This graduated, jacketed distillation receiver contains a vacuum manifold for removal of sample during operation without disturbing the system
Stopcocks on the manifold are 4mm bore; receiver stopcock is 3mm. Manifold has 10mm O.D. olive style connections. Fabricated from borosilicate glass.