Distillation Receiver, Vacuum Type, Ace Glass
Supplier: Ace Glass
6629-10
89055-728EA 483.33 USD
89055-728
Distillation Receiver, Vacuum Type, Ace Glass
Distillation Components Distillation Receivers
Receiver is graduated in 1mL subdivisions
Double scale, with glass stopcock. Fabricated from borosilicate glass.