Still, Solvent Recovery, Ace Glass
Supplier: Ace Glass
6614-30
89055-704EA 673.47 USD
89055-704
Still, Solvent Recovery, Ace Glass
Distillation Components Distillation Sets
When attached to the top of Snyder Column, this solvent recovery still is designed to recover solvent normally lost during Kuderna-Danish Evaporative Concentrations
The unit has a PTFE stopcock (1:5, 2mm bore) that provides the option for total reflux or takeoff. A #7 Ace-Thred on the side arm accepts 6-7mm O.D. tubing. A 89051-122 nylon bushing and FETFE o-ring are supplied with the solvent recovery adapter. Fabricated from borosilicate glass.
Ordering information: Complete item consists of 89051-454 Solvent Recovery Adapter with [ST] 24/40 joints and #7 Ace-Thred takeoff side arm (includes 89051-122 nylon bushing and FETFE o-ring; and 89053-782 Allihn Condenser (200mm jacket length, [ST] 24/40 joints. Optional items (not included): three 89059-876 PTFE sleeves; 89055-650 three-section Snyder Column; 89055-894 Flask (500mL); and 89055-920 graduated 10mL receiving flask. Solvent Recovery Adapter is also available with straight tube rather than #7 Ace-Thred joint (order 89051-456).