Distillation Head, Hennion Design, Ace Glass
Supplier: Ace Glass
6606-10
89055-700EA 1601.32 USD
89055-700
Distillation Head, Hennion Design, Ace Glass
Distillation Components Distillation Heads
This Hennion Design distillation head is used for variable reflux ratio, and can be operated under vacuum
The receiver can be removed without disturbing the system's vacuum. Fabricated from borosilicate glass.
Ordering information: This head comes complete with 89053-820 cold finger, which can also be ordered separately.