Distillation Head, Vacuum with Glass Stopcocks, Ace Glass
Supplier: Ace Glass
6613-12
89055-690EA 1710.68 USD
89055-690
Distillation Head, Vacuum with Glass Stopcocks, Ace Glass
Distillation Components Distillation Heads
This distillation head has a cold finger type condenser and 0-3mm PTFE stopcock for reflux control
The glass stopcocks on the manifold and reflux arms can be used to isolate the head from the receiver so that the receiver contents can be removed without disturbing the vacuum in the system. The head has a condenser on the takeoff arm and [ST] 10/30 thermometer joint designed for use with a 76mm immersion thermometer. The finger condenser is approximately 20.3cm long, with a [ST] 24/40 joint. The glass stopcocks are 3mm bore. Fabricated from borosilicate glass.