Distillation Apparatus, Markham Still, Ace Glass
Supplier: Ace Glass
6558-40
89055-622EA 1708.78 USD
89055-622
Distillation Apparatus, Markham Still, Ace Glass
Distillation Components Distillation Sets
This Markham Still distillation apparatus is used for volatile fatty acid tests
Fabricated from standard wall borosilicate glass tubing throughout. Hose connections on the still are for use with 3/8-inch I.D. tubing. Hose connections on the condenser are for use with 5/16-inch or 3/8-inch I.D. tubing.