Pressure Reactor Sparger Tubes, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
6444-04 6444-09 6444-16 6444-30 6444-38
89054-476EA 344.8 USD
89054-476 89054-478 89054-480 89054-482 89054-484
Pressure Reactor Sparger Tubes, PTFE, Ace Glass Incorporated
Tubes Test Tubes
This sparger tube is designed for use in #15 Ace-Thred with either one-piece or two-piece reactors.
  • O-ring on adapter is FETFE
  • Adapter and tube are fabricated from PTFE
  • Attach appropriate size tubing to top of adapter
Tube consists of a closed bottom, ¹/₂" Ext.Ø tube with a ¹/₈" Int.Ø bore. Bottom of tube has two ³/₆₄" holes, 90° apart; top has a ⁵/₁₆" male thread for connecting to bottom of #15 adapter. Adapter has a female thread in handle for tubing connection. Simply thread tube into bottom of #15 adapter. Thread into #15 Ace-Thred on reactor.
Ordering information: For replacement FETFE O-rings, see item 89060-984. Order tube and adapter separately.