One-Piece Unjacketed Pressure Reactor with Bottom Outlet System, Ace Glass Incorporated
Supplier: Ace Glass
6437-235 6437-238 6437-241 6437-244 6437-246
89054-414EA 13856.86 USD
89054-414 89054-416 89054-420 89054-422 89054-424
One-Piece Unjacketed Pressure Reactor with Bottom Outlet System, Ace Glass Incorporated
Condensers
Turn-key reactor system featuring a one-piece, unjacketed, pressure vessel with a combination Ace-thred valve and 28/15 o-ring ball joint bottom outlet
Vessel top has a 25 mm Ace-thred center neck and various multi-neck configurations. System is designed to perform positive pressure reactions at up to 45 psi at 100 °C. Stand Not Included.
Ordering information: Comes complete with (2) #15 PTFE Plugs, Condenser, Flask, Head, Manifold, Thermowell, Clamp, Funnel, (2) #15 PTFE Bushing, Bearing w/ Ace-threads, Stir Shaft, Flexible Shaft, PTFE Stirrer Blade, Krytox High Vacuum Grease, Swivel Coupling, Polypropylene Tubing, Motor Controller, Mounting Rod, PTFE Sealing Tape.