Unjacketed Filter Reactor, Pressure Rated to 35 psi, Ace Glass Incorporated
Supplier: Ace Glass
6384-05 6384-11 6384-115 6384-121 6384-125 6384-129 6384-133 6384-137 6384-15 6384-19 6384-225 6384-23 6384-231 6384-235 6384-239 6384-243 6384-247 6384-27
89054-088EA 641.22 USD
89054-088 89054-090 89054-092 89054-094 89054-096 89054-098 89054-100 89054-102 89054-104 89054-106 89054-108 89054-110 89054-112 89054-114 89054-116 89054-118 89054-120 89054-122
Unjacketed Filter Reactor, Pressure Rated to 35 psi, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
These heavy wall glass reactors are available with a 60mm (2.4"), 100mm (4"), or 150mm (6") diameter Duran style flange with O-Ring groove at top
Ordering Information: Complete item consists of reactor body with 60mm CAPFE O-Ring, head with Ace-Threds, clamp, PTFE bottom threaded adapter and valve with CAPFE O-Ring, retainer ring, 350µm polypropylene screen support, 100µm polyethylene support, adapter, bearing, stir rod, paddle agitator, condenser, addition funnel, and #7 Ace-Thred bushing. Size 80 bottom adapter also comes with a perforated glass plate support.
Connects to a head with a 7 quick release clamp. Reactor bottom has an Ace-Thred for installing a size 80 or size 50 adapter with a filter support and bottom drain valve. The 60mm reactor head has a #15 Ace-Thred center neck, (2) #15 and (1) #7 Ace-Thred side necks. The 100mm head has a #15 Ace-Thred center neck, (3) #15 and (1) #7 Ace-Thred side necks. The 150mm head has a #15 Ace-Thred center neck, (4) #15 and (1) #7 Ace-Thred side necks. The Ace-Threds allow for vacuum or pressure applications and for easy draining of material out of the reactor. The main bearing, funnel, and condenser are designed to connect to the threads on the head via compression bushing connections. The #7 Ace-Thred is used for connecting a ¼" O.D. tubing fitting to allow for a direct connection to a pressure source or a pressure manifold.