Desiccator Cabinets, Vertical Profile, Ace Glass
Supplier: Ace Glass
6256-43
89054-066EA 1861.42 USD
89054-066 89054-068
Desiccator Cabinets, Vertical Profile, Ace Glass
Desiccators Desiccator Cabinets
These stackable, plastic desiccator cabinets are perfect for cramped locations
Can be safely stacked up to two high. Cabinet offered in clear or blue with clear door and built-in hygrometer. Non-electric unit has three shelves, four shelf positions. Unique sealed construction — for air-tight, dust- and moisture-free storage. Two latches; provision for lock or tamper evident seal. Use any desiccant.