Condenser, Friedrichs, 250 mm, No. 3 Stopper Joint, Ace Glass Incorporated
Supplier: Ace Glass
5972-10
89053-890EA 422.13 USD
89053-890
Condenser, Friedrichs, 250 mm, No. 3 Stopper Joint, Ace Glass Incorporated
Condensers
Friedrichs condenser with molded spiral condensing surface and standard taper inner joint at bottom
Top tubulation is tooled to take a No. 3 stopper. Jacket length is 250mm. Use with 3/8-inch I.D. tubing, size D hose connections.