Syringe, Gas Tight, Luer-Lok, Ace Glass Incorporated
Supplier: Ace Glass
5932-08
89053-708EA 145.61 USD
89053-708
Syringe, Gas Tight, Luer-Lok, Ace Glass Incorporated
Syringes Syringes for Chromatography
Syringe with PTFE coated plunger and Luer-Lok
Used for sample introduction in chromatography solvent delivery systems. Has a capacity of 5mL.