Ion-Exchange Assembly, Laboratory Size, #25 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5881-502
89053-474EA 4417.39 USD
89053-474
Ion-Exchange Assembly, Laboratory Size, #25 Ace-Thred, Ace Glass Incorporated
Chromatography Columns
Unit is designed for metal recovery using ion-exchange resins such as amborane.
Ring stand has a rectangular steel base, and is finished in hard enamel, 127×229 mm. Rod is 9.5 mm Ext.Ø zinc-plated, 508 mm high.
Borosilicate glass column has #25 Ace-Thred at both ends. Polypropylene end fittings mate with Ace-Threds and form a leak-tight seal with FETFE o-rings. Each end fitting has a 'coin' tightened retainer that holds packing support screen firmly in place; screen can be replaced in seconds. Fittings have 1/4-inch npt female thread for connecting 3/8-inch Ext.Ø flexible vinyl tubing from pump and reservoir using a 1/4-inch to 3/8-inch tubing connector. Normal flow is in upward direction. Column is easily held by ring stand and clamp.
Ordering information: Complete unit consists of one each of column 89052-340, pump 89077-588, tubing 89077-036, tubing 89077-066, filter 89052-200, filter 89052-210, O-ring 89061-004, ring stand 89053-472, clamp 89083-210 and two each of adapter 89052-994, connector 89077-404, and connector 89077-418.