Column Extenders, Ace Glass Incorporated
Supplier: Ace Glass
5862-72 5862-73 5862-74 5862-77 5862-78 5862-79
89053-092EA 589.48 USD
89053-092 89053-094 89053-096 89053-098 89053-100 89053-102
Column Extenders, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These extenders can be added to columns like 89053-076, 89053-084, or 89053-058 (and others) to increase column height
The extenders have an O-Ring grooved Duran flat flange on one end that attaches to column heads like 89053-052, 89053-054 or 89053-056, and a ground flat flange at the other end that attaches to the column. Fabricated from borosilicate glass.
Ordering information: Extenders are supplied with one silicone O-Ring. For replacement silicone O-Rings, order 71000-162 (4") or 71000-164 (4"). For replacement CAPFE O-Rings, order 89061-166 (for 4") or 89061-168 (for 4"). For clamps (not included), order 89055-110 (4") or 89055-112 (6") quick-release clamp.