Adapter, Luer, Ace Glass Incorporated
Supplier: Ace Glass
5852-10
89052-932PK 78.28 USD
89052-932
Adapter, Luer, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter consists of thin wall biological grade vinyl tubing that connects to an outer Luer taper and an inner Luer-Lok taper.
  • Constructed of thin wall tubing
  • One outer Luer taper and one inner Luer-Lok taper
  • Bottom drip adapter
Adapter is constructed of 84 cm of thin wall biological grade vinyl tubing. Tubing is approximately 2 mm I.D., and connects to a bottom drip adapter.